missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-(2-Aminoethyl)-1-methylpyrrolidine, 97%, ACROS Organics™

Manufacturer:  Acros Organics AC160860010

 View more versions of this product

Catalog No. AC160860010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

2-(2-Aminoethyl)-1-methylpyrrolidine
Authentic
1.4670 to 1.4690 (20°C, 589nm)
1g
96.0
97%
MFCD00003175
PNHGJPJOMCXSKN-UHFFFAOYSA-N
2-(1-methylpyrrolidin-2-yl)ethanamine
98388
Powder
0.8800g/mL
Glass bottle
Beige to Brown
51387-90-7
100.0
C7H16N2
2-2-aminoethyl-1-methylpyrrolidine, 2-1-methylpyrrolidin-2-yl ethanamine, 2-pyrrolidineethanamine, 1-methyl, 2-2'-aminoethyl-1-n-methylpyrrolidine, n-methyl-2-2-aminoethyl pyrrolidine, 1-methyl-2-2-aminoethyl pyrrolidine, 2-1-methylpyrrolidin-2-yl ethan-1-amine, 2-1-methyl-2-pyrrolidinyl ethylamine, 1-methyl-2-pyrrolidineethanamine, 1-methylpyrrolidine-2-ethylamine
CN1CCCC1CCN
128.219
128.22
≥96.0% (GC)

Chemical Identifiers

51387-90-7
128.219
PNHGJPJOMCXSKN-UHFFFAOYSA-N
98388
CN1CCCC1CCN
C7H16N2
MFCD00003175
2-2-aminoethyl-1-methylpyrrolidine, 2-1-methylpyrrolidin-2-yl ethanamine, 2-pyrrolidineethanamine, 1-methyl, 2-2'-aminoethyl-1-n-methylpyrrolidine, n-methyl-2-2-aminoethyl pyrrolidine, 1-methyl-2-2-aminoethyl pyrrolidine, 2-1-methylpyrrolidin-2-yl ethan-1-amine, 2-1-methyl-2-pyrrolidinyl ethylamine, 1-methyl-2-pyrrolidineethanamine, 1-methylpyrrolidine-2-ethylamine
2-(1-methylpyrrolidin-2-yl)ethanamine
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage. Harmful if swallowed.

GHS P Statement
IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

Danger

SDS Documents