missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-(2-Aminoethyl)-1-methylpyrrolidine, 97%, Thermo Scientific™
Chemical Identifiers
CAS | 51387-90-7 |
---|---|
Molecular Formula | C7H16N2 |
Molecular Weight (g/mol) | 128.22 |
MDL Number | MFCD00003175 |
InChI Key | PNHGJPJOMCXSKN-UHFFFAOYNA-N |
Synonym | 2-2-aminoethyl-1-methylpyrrolidine, 2-1-methylpyrrolidin-2-yl ethanamine, 2-pyrrolidineethanamine, 1-methyl, 2-2'-aminoethyl-1-n-methylpyrrolidine, n-methyl-2-2-aminoethyl pyrrolidine, 1-methyl-2-2-aminoethyl pyrrolidine, 2-1-methylpyrrolidin-2-yl ethan-1-amine, 2-1-methyl-2-pyrrolidinyl ethylamine, 1-methyl-2-pyrrolidineethanamine, 1-methylpyrrolidine-2-ethylamine |
PubChem CID | 98388 |
IUPAC Name | 2-(1-methylpyrrolidin-2-yl)ethanamine |
SMILES | CN1CCCC1CCN |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC160860050
|
Thermo Scientific Chemicals
160860050 |
5g | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
51387-90-7 | |
128.22 | |
PNHGJPJOMCXSKN-UHFFFAOYNA-N | |
98388 | |
CN1CCCC1CCN |
C7H16N2 | |
MFCD00003175 | |
2-2-aminoethyl-1-methylpyrrolidine, 2-1-methylpyrrolidin-2-yl ethanamine, 2-pyrrolidineethanamine, 1-methyl, 2-2'-aminoethyl-1-n-methylpyrrolidine, n-methyl-2-2-aminoethyl pyrrolidine, 1-methyl-2-2-aminoethyl pyrrolidine, 2-1-methylpyrrolidin-2-yl ethan-1-amine, 2-1-methyl-2-pyrrolidinyl ethylamine, 1-methyl-2-pyrrolidineethanamine, 1-methylpyrrolidine-2-ethylamine | |
2-(1-methylpyrrolidin-2-yl)ethanamine |
Specifications
51387-90-7 | |
100.0 | |
65°C | |
96% min. (GC) | |
C7H16N2 | |
MFCD00003175 | |
0.88 | |
PNHGJPJOMCXSKN-UHFFFAOYNA-N | |
2-(1-methylpyrrolidin-2-yl)ethanamine | |
98388 | |
97% |
96.0 | |
0.8800g/mL | |
Authentic | |
Glass bottle | |
1.4684 | |
5g | |
2-2-aminoethyl-1-methylpyrrolidine, 2-1-methylpyrrolidin-2-yl ethanamine, 2-pyrrolidineethanamine, 1-methyl, 2-2'-aminoethyl-1-n-methylpyrrolidine, n-methyl-2-2-aminoethyl pyrrolidine, 1-methyl-2-2-aminoethyl pyrrolidine, 2-1-methylpyrrolidin-2-yl ethan-1-amine, 2-1-methyl-2-pyrrolidinyl ethylamine, 1-methyl-2-pyrrolidineethanamine, 1-methylpyrrolidine-2-ethylamine | |
CN1CCCC1CCN | |
128.22 | |
128.22 | |
2-(2-Aminoethyl)-1-methylpyrrolidine |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
GHS Signal Word: Danger
EINECSNumber : 257-169-
RUO â Research Use Only