missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(2-Amino-5-nitrophenyl)-5-methyl-4-phenylthiazole, 97%

Chemical Identifiers

CAS 1965305-02-5
Molecular Formula C16H13N3O2S
Molecular Weight (g/mol) 311.359
InChI Key UCBKPBJMEXATSY-UHFFFAOYSA-N
Synonym 2-2-amino-5-nitrophenyl-5-methyl-4-phenylthiazole, 2-5-methyl-4-phenyl-1,3-thiazol-2-yl-4-nitroaniline
PubChem CID 86277505
IUPAC Name 2-(5-methyl-4-phenyl-1,3-thiazol-2-yl)-4-nitroaniline
SMILES CC1=C(N=C(S1)C2=C(C=CC(=C2)[N+](=O)[O-])N)C3=CC=CC=C3
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH66305MD View Documents Alfa Aesar™
H66305MD
250mg N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAH6630503 View Documents Alfa Aesar™
H6630503
1g N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

1965305-02-5
311.359
2-2-amino-5-nitrophenyl-5-methyl-4-phenylthiazole, 2-5-methyl-4-phenyl-1,3-thiazol-2-yl-4-nitroaniline
2-(5-methyl-4-phenyl-1,3-thiazol-2-yl)-4-nitroaniline
C16H13N3O2S
UCBKPBJMEXATSY-UHFFFAOYSA-N
86277505
CC1=C(N=C(S1)C2=C(C=CC(=C2)[N+](=O)[O-])N)C3=CC=CC=C3

Specifications

Yellow
1965305-02-5
2-2-amino-5-nitrophenyl-5-methyl-4-phenylthiazole, 2-5-methyl-4-phenyl-1,3-thiazol-2-yl-4-nitroaniline
CC1=C(N=C(S1)C2=C(C=CC(=C2)[N+](=O)[O-])N)C3=CC=CC=C3
311.359
311.36
97%
250mg
C16H13N3O2S
UCBKPBJMEXATSY-UHFFFAOYSA-N
2-(5-methyl-4-phenyl-1,3-thiazol-2-yl)-4-nitroaniline
86277505
Crystalline Solid
2-(2-Amino-5-nitrophenyl)-5-methyl-4-phenylthiazole