Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(2-Amino-5-nitrophenyl)-4-phenylthiazole, 97%

Chemical Identifiers

Chemical Name or Material 2-(2-Amino-5-nitrophenyl)-4-phenylthiazole
CAS 41450-34-4
Formula Weight 297.33
Synonym 4-Nitro-2-(4-phenyl-2-thiazolyl)aniline
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6647803

 
alfa aesar™
H6647803
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH66478MD

 
alfa aesar™
H66478MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Melting Point 188° to 189°C
Formula Weight 297.33
CAS 41450-34-4
Chemical Name or Material 2-(2-Amino-5-nitrophenyl)-4-phenylthiazole
Assay 97%
Synonym 4-Nitro-2-(4-phenyl-2-thiazolyl)aniline
Color Yellow
Physical Form Crystalline Solid
Quantity 1g