missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,2′,7,7′-Tetrabromo-9,9′-spirobi[9H-fluorene], TCI America™

Manufacturer:  TCI America T25975G

 View more versions of this product

Catalog No. T25975G



Specifications

Specifications

2,2′,7,7′-Tetrabromo-9,9′-spirobi[9H-fluorene]
5g
C25H12Br4
MASXXNUEJVMYML-UHFFFAOYSA-N
2,2',7,7'-tetrabromo-9,9'-spirobi[fluorene]
371282
Crystal-Powder at 20°C
White-Yellow
128055-74-3
MFCD08704220
C1=CC2=C(C=C1Br)C3(C4=C2C=CC(=C4)Br)C5=C(C=CC(=C5)Br)C6=C3C=C(C=C6)Br
631.987
631.99
≥98.0% (T)

Chemical Identifiers

128055-74-3
631.987
MASXXNUEJVMYML-UHFFFAOYSA-N
2,2',7,7'-tetrabromo-9,9'-spirobi[fluorene]
C25H12Br4
MFCD08704220
371282
C1=CC2=C(C=C1Br)C3(C4=C2C=CC(=C4)Br)C5=C(C=CC(=C5)Br)C6=C3C=C(C=C6)Br
Safety and Handling

Safety and Handling

TSCA : No

Documents