missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2,2',7,7'-Tetrabromo-9,9'-spirobi[9H-fluorene] 98.0+%, TCI America™
Supplier: TCI America T25971G
Specifications
2,2′,7,7′-Tetrabromo-9,9′-spirobi[9H-fluorene] | |
White-Yellow | |
MFCD08704220 | |
MASXXNUEJVMYML-UHFFFAOYSA-N | |
2,2',7,7'-tetrabromo-9,9'-spirobi[fluorene] | |
371282 | |
≥98.0% (T) |
128055-74-3 | |
C25H12Br4 | |
1 g | |
BrC1=CC2=C(C=C1)C1=C(C=C(Br)C=C1)C21C2=C(C=CC(Br)=C2)C2=C1C=C(Br)C=C2 | |
631.99 | |
631.99 | |
Crystalline Powder |
Chemical Identifiers
128055-74-3 | |
631.99 | |
MASXXNUEJVMYML-UHFFFAOYSA-N | |
2,2',7,7'-tetrabromo-9,9'-spirobi[fluorene] |
C25H12Br4 | |
MFCD08704220 | |
371282 | |
BrC1=CC2=C(C=C1)C1=C(C=C(Br)C=C1)C21C2=C(C=CC(Br)=C2)C2=C1C=C(Br)C=C2 |
Safety and Handling
TSCA : No