missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(2,6-Dichlorophenyl)thioacetamide, 97%

$96.97 - $240.12

Chemical Identifiers

CAS 17518-49-9
Molecular Formula C8H7Cl2NS
Formula Weight 220.12
MDL Number MFCD00052483
Chemical Name or Material 2-(2,6-Dichlorophenyl)thioacetamide
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH2674603 SDS Alfa Aesar™
H2674603
1g Each for $240.12
AAH26746MD SDS Alfa Aesar™
H26746MD
250mg Each for $96.97
Specifications Description & Specifications

Chemical Identifiers

17518-49-9
220.12
2-(2,6-Dichlorophenyl)thioacetamide
C8H7Cl2NS
MFCD00052483

Specifications

1g
C8H7Cl2NS
MFCD00052483
2-(2,6-Dichlorophenyl)thioacetamide
17518-49-9
220.12
189° to 191°C
97%

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger