Please login to your online account to display your discounted pricing

2-(2,6-Dichlorophenyl)ethanimidamide hydrochloride, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material 2-(2, 6-Dichlorophenyl)ethanimidamide hydrochloride
CAS 175276-76-3
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C8H8Cl2N2·ClH
Linear Formula Cl2C6H3CH2C(=NH)NH2·HCl
Formula Weight 239.53
MDL Number MFCD00053014
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

SPB00311EA

  SDS
fisher scientific company
SPB00311EA
Amber Glass Bottle 10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 239.53
Quantity 10g
Assay 97%
CAS Min % 97.0
Assay Percent Range 97%
Linear Formula Cl2C6H3CH2C(=NH)NH2·HCl
CAS 175276-76-3
Health Hazard 1 GHS Signal Word: Warning
Molecular Formula C8H8Cl2N2·ClH
MDL Number MFCD00053014
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material 2-(2, 6-Dichlorophenyl)ethanimidamide hydrochloride
Packaging Amber Glass Bottle
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Harmful if inhaled.
Harmful in contact with skin.
May cause respiratory irritation.
Causes skin irritation.
Harmful if swallowed.