Learn More
2,2,6,6-Tetramethylpiperidinooxy, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 200380050
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagentSpecifications
2,2,6,6-Tetramethylpiperidinooxy | |
2564-83-2 | |
100.0 | |
Yellow to Green | |
Authentic | |
Glass bottle | |
MFCD00009599 | |
16,324; 17,332 | |
TEMPO | |
QYTDEUPAUMOIOP-UHFFFAOYSA-N | |
(2,2,6,6-tetramethylpiperidin-1-yl)oxidanyl | |
156.25 | |
Powder |
98% | |
97.5 | |
36°C to 40°C | |
67°C | |
97.5 % min. (GC) | |
C9H18NO | |
5 g | |
15, 9283 | |
Solubility in water: 151mg/l (20°C) | |
CC1(C)CCCC(C)(C)N1[O] | |
156.25 | |
98% |
Chemical Identifiers
2564-83-2 | |
156.25 | |
QYTDEUPAUMOIOP-UHFFFAOYSA-N | |
(2,2,6,6-tetramethylpiperidin-1-yl)oxidanyl |
C9H18NO | |
MFCD00009599 | |
TEMPO | |
CC1(C)CCCC(C)(C)N1[O] |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Harmful to aquatic life with long lasting effects.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat
GHS Signal Word: Danger
EINECSNumber : 219-888-8
RTECSNumber : TN8991900
TSCA : TSCA
RUO – Research Use Only