Learn More
2,2,6,6-Tetramethylpiperidine, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 151950100
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
2,2,6,6-Tetramethylpiperidine | |
Colorless to Yellow | |
152.0°C | |
Authentic | |
Glass bottle | |
1.4430 to 1.4460 | |
20,129 | |
norpempidine, piperidine, 2,2,6,6-tetramethyl, 2,2,6,6-tetramethyl piperidine, unii-44n9s1ycfm, 2,2,6,6-tetramethylpeperidine, 44n9s1ycfm, 2,2,6,6 tetramethyl piperidine, 2,2,6,6-tetramethyl-piperidine, pubchem7259, acmc-209p6i | |
RKMGAJGJIURJSJ-UHFFFAOYSA-N | |
2,2,6,6-tetramethylpiperidine | |
13035 | |
98% |
768-66-1 | |
0.8300g/mL | |
24°C | |
97.5% min. (GC) | |
C9H19N | |
10 g | |
0.83 | |
Solubility in water: miscible. Other solubilities: soluble in toluene, dimethylformamide | |
CC1(CCCC(N1)(C)C)C | |
141.26 | |
141.26 | |
Liquid |
Chemical Identifiers
768-66-1 | |
141.26 | |
norpempidine, piperidine, 2,2,6,6-tetramethyl, 2,2,6,6-tetramethyl piperidine, unii-44n9s1ycfm, 2,2,6,6-tetramethylpeperidine, 44n9s1ycfm, 2,2,6,6 tetramethyl piperidine, 2,2,6,6-tetramethyl-piperidine, pubchem7259, acmc-209p6i | |
2,2,6,6-tetramethylpiperidine |
C9H19N | |
RKMGAJGJIURJSJ-UHFFFAOYSA-N | |
13035 | |
CC1(CCCC(N1)(C)C)C |
Safety and Handling
GHS H Statement
Flammable liquid and vapor.
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective glo
GHS Signal Word: Danger
EINECSNumber : 212-199-3
RUO – Research Use Only