missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,2,6,6-Tetramethylpiperidine 1-Oxyl Free Radical, TCI America™

Manufacturer:  TCI America T156025G

 View more versions of this product

Catalog No. T156025G



Specifications

Specifications

2,2,6,6-Tetramethylpiperidine 1-Oxyl Free Radical
39°C
2564-83-2
MFCD00009599
TEMPO
Crystal-Powder at 20°C
Undesignated
25g
C9H18NO
3263
156.25
≥98.0% (GC,T)

Chemical Identifiers

2564-83-2
C9H18NO
MFCD00009599
TEMPO
Safety and Handling

Safety and Handling

RTECSNumber : TN8991900

TSCA : Yes

Recommended Storage : Refrigerator

Documents