Please login to your online account to display your discounted pricing

2,2,6,6-Tetramethylpiperidine-1-oxyl-4-amino-4-carboxylic acid, 96%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2,2,6,6-Tetramethylpiperidine-1-oxyl-4-amino-4-carboxylic acid
Name Note 96%
CAS 15871-57-5
CAS Min % 95.0
CAS Max % 100.0
Assay ≥95% (GC)
Assay Percent Range 96%
Molecular Formula C10H19N2O3
Formula Weight 215.27
MDL Number MFCD00800575
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC301452500

  SDS
fisher scientific acros
301452500
Glass bottle 250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC301450010

  SDS
thermo fisher scientific
AC301450010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥95% (GC)
Physical Form Crystalline Powder
Formula Weight 215.27
Quantity 250mg
Color White to off-white
TSCA TSCA
CAS Max % 100.0
CAS Min % 95.0
Synonym 4-Amino-4-carboxy-2, 2, 6
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Glass bottle
Infrared Spectrum Authentic
Molecular Formula C10H19N2O3
Assay Percent Range 96%
Name Note 96%
Chemical Name or Material 2,2,6,6-Tetramethylpiperidine-1-oxyl-4-amino-4-carboxylic acid
MDL Number MFCD00800575
CAS 15871-57-5
Safety and Handling

Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.