missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,2,6,6-Tetramethylpiperidine-1-oxyl-4-amino-4-carboxylic acid, 96%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC301450010

 View more versions of this product

Catalog No. AC301450010

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

≥95% (GC)
215.27
White to off-white
100.0
4-Amino-4-carboxy-2, 2, 6
2,2,6,6-Tetramethylpiperidine-1-oxyl-4-amino-4-carboxylic acid
96%
C10H19N2O3
Crystalline Powder
1g
95.0
Glass bottle
Authentic
96%
15871-57-5
MFCD00800575

Chemical Identifiers

15871-57-5
100.0
C10H19N2O3
MFCD00800575
2,2,6,6-Tetramethylpiperidine-1-oxyl-4-amino-4-carboxylic acid, 96%
95.0
96%
215.27
4-Amino-4-carboxy-2, 2, 6
≥95% (GC)

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

TSCA : TSCA