missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,2,6,6-Tetramethyl-4-piperidyl Methacrylate, TCI America™

Manufacturer:  TCI America T2324500G

 View more versions of this product

Catalog No. T2324500G



Specifications

Specifications

2,2,6,6-Tetramethyl-4-piperidyl Methacrylate
61°C
31582-45-3
Methacrylic Acid 2,2,6,6-Tetramethyl-4-piperidyl Ester
CC(=C)C(=O)OC1CC(NC(C1)(C)C)(C)C
225.332
225.33
≥98.0% (GC)
White-Yellow
500g
C13H23NO2
UFLXKQBCEYNCDU-UHFFFAOYSA-N
(2,2,6,6-tetramethylpiperidin-4-yl) 2-methylprop-2-enoate
9815965
Crystal-Powder at 20°C

Chemical Identifiers

31582-45-3
225.332
Methacrylic Acid 2,2,6,6-Tetramethyl-4-piperidyl Ester
(2,2,6,6-tetramethylpiperidin-4-yl) 2-methylprop-2-enoate
C13H23NO2
UFLXKQBCEYNCDU-UHFFFAOYSA-N
9815965
CC(=C)C(=O)OC1CC(NC(C1)(C)C)(C)C
Safety and Handling

Safety and Handling

EINECSNumber : (5)-6022

TSCA : No

Recommended Storage : Refrigerator

Documents