Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,2,6,6-Tetramethyl-4-piperidone monohydrate, 99%

$39.03 - $650.92

Chemical Identifiers

Chemical Name or Material 2,2,6,6-Tetramethyl-4-piperidone monohydrate
CAS 10581-38-1
Molecular Formula C9H17NO·H2O
Formula Weight 173.26 (155.24 Anhydrous)
MDL Number MFCD00149393
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1621109

 
alfa aesar™
A1621109
10g Each for $39.03

AAA1621118

 
alfa aesar™
A1621118
50g Each for $150.68

AAA1621130

 
alfa aesar™
A1621130
250g Each for $650.92
Description & Specifications

Specifications

Formula Weight 173.26 (155.24 Anhydrous)
Chemical Name or Material 2,2,6,6-Tetramethyl-4-piperidone monohydrate
TSCA TSCA
Assay 99%
Molecular Formula C9H17NO·H2O
MDL Number MFCD00149393
CAS 10581-38-1
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 63° to 61°C
Quantity 10g
Recommended Storage 2° to 8°C
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

2° to 8°C