missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,2,6,6-Tetramethyl-3,5-heptanedione, 98%, ACROS Organics™

Manufacturer:  Acros Organics 168320250

 View more versions of this product

Catalog No. AC168320250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Acylation reagent
Specifications

Specifications

2,2,6,6-Tetramethyl-3,5-heptanedione
67°C
Glass bottle
0.88
Colorless to Yellow
1118-71-4
C11H20O2
dipivaloylmethane, 2,2,6,6-tetramethyl-3,5-heptanedione, 3,5-heptanedione, 2,2,6,6-tetramethyl, unii-r8ui909hoy, tmhd, 2,2,6,6-tetramethyl-3,5-heptanedione dipivaloylmethane, r8ui909hoy, 2,2,6,6-tetramethyl-heptane-3,5-dione, 2,2,6,6-tetramethyl-3,5-heptanedione dipivaloylmethane, pubchem12497
YRAJNWYBUCUFBD-UHFFFAOYSA-N
2,2,6,6-tetramethylheptane-3,5-dione
70700
Liquid
0.8800g/mL
Authentic
1.4579 to 1.4599
72.0°C to 73.0°C (6.0mmHg)
25g
98%
(CH3)3CCOCH2COC(CH3)3
Solubility in water: insoluble.
CC(C)(C)C(=O)CC(=O)C(C)(C)C
184.279
184.28
98%

Chemical Identifiers

1118-71-4
184.279
dipivaloylmethane, 2,2,6,6-tetramethyl-3,5-heptanedione, 3,5-heptanedione, 2,2,6,6-tetramethyl, unii-r8ui909hoy, tmhd, 2,2,6,6-tetramethyl-3,5-heptanedione dipivaloylmethane, r8ui909hoy, 2,2,6,6-tetramethyl-heptane-3,5-dione, 2,2,6,6-tetramethyl-3,5-heptanedione dipivaloylmethane, pubchem12497
2,2,6,6-tetramethylheptane-3,5-dione
C11H20O2
YRAJNWYBUCUFBD-UHFFFAOYSA-N
70700
CC(C)(C)C(=O)CC(=O)C(C)(C)C
SDS Documents