Please login to your online account to display your discounted pricing

2,2,6,6-Tetramethyl-3,5-heptanedione, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2,2,6,6-Tetramethyl-3,5-heptanedione
Assay ≥97.5% (GC)
Assay Percent Range 98%
Molecular Formula C11H20O2
Linear Formula (CH3)3CCOCH2COC(CH3)3
Formula Weight 184.28
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC168320050

  SDS
fisher scientific acros
168320050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC168320250

  SDS
fisher scientific acros
168320250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC168321000

  SDS
thermo fisher scientific
AC168321000
Glass bottle 100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5% (GC)
Physical Form Liquid
Formula Weight 184.28
Quantity 5g
Color Clear colorless to slightly yellow
Density 0.88g/mL
Packaging Glass bottle
Molecular Formula C11H20O2
CAS 1118-71-4
Chemical Name or Material 2,2,6,6-Tetramethyl-3,5-heptanedione
Infrared Spectrum Authentic
Assay Percent Range 98%
Linear Formula (CH3)3CCOCH2COC(CH3)3
Refractive Index 1.4579 to 1.4599 (20°C, 589nm)

Acylation reagent