Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,2,6,6-Tetramethyl-3,5-heptanedione, 98%

$189.85 - $631.34

Chemical Identifiers

Chemical Name or Material 2,2,6,6-Tetramethyl-3,5-heptanedione
CAS 1118-71-4
Molecular Formula C11H20O2
Formula Weight 184.28
MDL Number MFCD00008848
Beilstein 1447269
Synonym Dipivaloylmethane; Dpm-H
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1511814

  SDS
alfa aesar™
A1511814
25g Each for $189.85

AAA1511822

  SDS
alfa aesar™
A1511822
100g Each for $631.34
Description & Specifications

Specifications

Formula Weight 184.28
Quantity 25g
MDL Number MFCD00008848
Boiling Point 72° to 73°C (6mmHg)
Molecular Formula C11H20O2
Health Hazard 2 GHS H Statement
H302-H227
Harmful if swallowed.
Combustible liquid.
Flash Point 67°C (152°F)
Beilstein 1447269
Chemical Name or Material 2,2,6,6-Tetramethyl-3,5-heptanedione
Synonym Dipivaloylmethane; Dpm-H
TSCA TSCA
Assay 98%
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P210-P280-P264-P301+P312-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Wash thoroughly after handling.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
CAS 1118-71-4

Applications
2,2,6,6-Tetramethyl-3,5-heptanedione is used in the synthesis of α-aryl-β-diketones1 and dicyanamidobenzene-bridge diruthenium complex.

Solubility
Difficult to mix.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Safety and Handling

Warning

GHS P Statement
P210-P280-P264-P301+P312-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Wash thoroughly after handling.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H227
Harmful if swallowed.
Combustible liquid.