Learn More
2,2':6',2''-Terpyridine, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1775003
Description
2,2';6',2'-terpyridine is widely utilized in the field of supramolecular chemistry, which is used to manufacture the racks, ladders and grids, helicates, catenanes and dendrimers. It plays an important role as a ligand in coordination chemistry. Its complexes are employed in the oxidation of alcohols, the carbonylation of aromatic compounds and as oxygen-binding molecules. Functionalized terpyridine ligands were used in semiconductors and solar panels. It forms chealate complexes with europium(III) and terbium(III), which is used in protein labelling.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2,2′;6′,2″-terpyridine is widely utilized in the field of supramolecular chemistry, which is used to manufacture the racks, ladders and grids, helicates, catenanes and dendrimers. It plays an important role as a ligand in coordination chemistry. Its complexes are employed in the oxidation of alcohols, the carbonylation of aromatic compounds and as oxygen-binding molecules. Functionalized terpyridine ligands were used in semiconductors and solar panels. It forms chealate complexes with europium(III) and terbium(III), which is used in protein labelling.
Solubility
Soluble in most organic solvents.
Notes
Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
Specifications
2, 2':6', 2''-Terpyridine | |
89°C to 92°C | |
C15H11N3 | |
1 g | |
2,2':6',2-terpyridine, tripyridyl, tripyridine, 2,6-bis 2-pyridyl pyridine, terpy, terpyridine, 2,2',2-terpyridine, 2,2',2-terpyridyl, 2,2',2-tripyridyl, 2,2',2-tripyridine | |
DRGAZIDRYFYHIJ-UHFFFAOYSA-N | |
2,6-dipyridin-2-ylpyridine | |
70848 | |
233.28 |
1148-79-4 | |
0.97 | |
MFCD00006213 | |
UN2811 | |
Soluble in most organic solvents. | |
C1=CC=C(N=C1)C1=CC=CC(=N1)C1=CC=CC=N1 | |
233.27 | |
CHEBI:245199 | |
97% |
Chemical Identifiers
1148-79-4 | |
233.27 | |
DRGAZIDRYFYHIJ-UHFFFAOYSA-N | |
70848 | |
2,6-dipyridin-2-ylpyridine |
C15H11N3 | |
MFCD00006213 | |
2,2':6',2-terpyridine, tripyridyl, tripyridine, 2,6-bis 2-pyridyl pyridine, terpy, terpyridine, 2,2',2-terpyridine, 2,2',2-terpyridyl, 2,2',2-tripyridyl, 2,2',2-tripyridine | |
CHEBI:245199 | |
C1=CC=C(N=C1)C1=CC=CC(=N1)C1=CC=CC=N1 |
Safety and Handling
GHS H Statement
H300-H310-H315-H319-H335
P261-P262-P264b-P270-P271-P280-P301+P310-P302+P350-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P361-P363-P501c
H300+H310-H315-H319-H335
DOTInformation : Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber : 214-559-5
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only