missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,2':6',2''-Terpyridine-4'-carboxylic acid, 95%

Manufacturer:  Alfa Aesar™ H37760MD

 View more versions of this product

Catalog No. AAH37760MD


 

Description

Description

Applications
2,2′:6′,2′′-Terpyridine-4′-carboxylic acid is used as dyestuff intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications

Specifications

2,2':6',2''-Terpyridine-4'-carboxylic acid
95%
MFCD04114290
ZYTWXMBGOUJDHJ-UHFFFAOYSA-N
2,6-dipyridin-2-ylpyridine-4-carboxylic acid
2762749
Powder
280°C to 282°C
148332-36-9
C16H11N3O2
4'-Carboxy-2,2':6',2''-terpyridine
C1=CC=NC(=C1)C2=CC(=CC(=N2)C3=CC=CC=N3)C(=O)O
277.283
277.28
White through Cream
250mg

Chemical Identifiers

148332-36-9
277.283
ZYTWXMBGOUJDHJ-UHFFFAOYSA-N
2762749
C16H11N3O2
MFCD04114290
4'-Carboxy-2,2':6',2''-terpyridine
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

SDS Documents