Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,2,5-Trimethyl-3-hexen-1-ol, 97%

Chemical Identifiers

Chemical Name or Material 2,2,5-Trimethyl-3-hexen-1-ol
Molecular Formula C9H18O
Formula Weight 142.24
MDL Number MFCD03788753
Beilstein 6774184
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1970203

  SDS
alfa aesar™
L1970203
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1970206

  SDS
alfa aesar™
L1970206
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 142.24
Chemical Name or Material 2,2,5-Trimethyl-3-hexen-1-ol
Assay 97%
Molecular Formula C9H18O
Beilstein 6774184
Quantity 1g
MDL Number MFCD03788753