Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(2,5-Dimethoxyphenylamino)ethanol

$263.26 - $263.26

Chemical Identifiers

Chemical Name or Material 2-(2,5-Dimethoxyphenylamino)ethanol
CAS 28226-20-2
Molecular Formula C10H15NO3
Formula Weight 197.24g/mol
MDL Number MFCD03701227
Synonym N-(2-Hydroxyethyl)-2,5-dimethoxyaniline
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH51177MD

 
alfa aesar™
H51177-MD
250mg Each for $263.26
Description & Specifications

Specifications

Quantity 250mg
Quantity 250mg
Chemical Name or Material 2-(2,5-Dimethoxyphenylamino)ethanol
CAS 28226-20-2
Molecular Formula C10H15NO3
Formula Weight 197.24g/mol
MDL Number MFCD03701227
Synonym N-(2-Hydroxyethyl)-2,5-dimethoxyaniline
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.