Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(2,5-Difluorophenyl)ethanol, 97%

$56.50 - $187.80

Chemical Identifiers

Chemical Name or Material 2-(2,5-Difluorophenyl)ethanol
CAS 862255-56-9
Molecular Formula C8H8F2O
Formula Weight 158.15
MDL Number MFCD00142840
Synonym 2,5-Difluorophenethyl alcohol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3208203

  SDS
alfa aesar™
H3208203
1g Each for $187.80

AAH32082MD

  SDS
alfa aesar™
H32082MD
250mg Each for $56.50
Description & Specifications

Specifications

Formula Weight 158.15
Assay 97%
MDL Number MFCD00142840
CAS 862255-56-9
Chemical Name or Material 2-(2,5-Difluorophenyl)ethanol
Molecular Formula C8H8F2O
Synonym 2,5-Difluorophenethyl alcohol
Quantity 1g