Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,2',5,5'-Tetramethylbiphenyl, 98%

$91.73 - $338.64

Chemical Identifiers

Chemical Name or Material 2,2',5,5'-Tetramethylbiphenyl
CAS 3075-84-1
Molecular Formula C16H18
Formula Weight 210.32
MDL Number MFCD00151846
Beilstein 2046200
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1333003

 
alfa aesar™
L1333003
1g Each for $91.73

AAL1333006

 
alfa aesar™
L1333006
5g Each for $338.64
Description & Specifications

Specifications

Formula Weight 210.32
Chemical Name or Material 2,2',5,5'-Tetramethylbiphenyl
Molecular Formula C16H18
MDL Number MFCD00151846
CAS 3075-84-1
Assay 98%
Beilstein 2046200
Boiling Point 284°C (732mmHg)
Melting Point 48° to 51°C
Quantity 1g

Applications
2,2′,5,5′-Tetramethylbiphenyl is used to make other chemicals and as an intermediate in pharmaceuticals.

Solubility
Insoluble in water.

Notes
Incompatible with strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.