missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2,2',5,5'-Tetramethylbiphenyl, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L1333006
Description
2,2',5,5'-Tetramethylbiphenyl is used to make other chemicals and as an intermediate in pharmaceuticals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2,2′,5,5′-Tetramethylbiphenyl is used to make other chemicals and as an intermediate in pharmaceuticals.
Solubility
Insoluble in water.
Notes
Incompatible with strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
2,2',5,5'-Tetramethylbiphenyl | |
3075-84-1 | |
5 g | |
MFCD00151846 | |
2,2',5,5'-tetramethylbiphenyl, 2,2',5,5'-tetramethyl-1,1'-biphenyl, 1,1'-biphenyl, 2,2',5,5'-tetramethyl, biphenyl-, 2,2',5,5'-tetramethyl, 2-2,5-dimethylphenyl-1,4-dimethylbenzene, acmc-20aokf, 2,5,2',5'-tetramethylbiphenyl, 1,1'-biphenyl,2,2',5,5'-tetramethyl, 2,2',5,5'-tetramethyl-1,1'-biphenyl # | |
ZHTROMYSDSTCCE-UHFFFAOYSA-N | |
2-(2,5-dimethylphenyl)-1,4-dimethylbenzene | |
137818 | |
98% |
48°C to 51°C | |
284°C (732 mmHg) | |
C16H18 | |
2046200 | |
Insoluble in water. | |
CC1=CC=C(C)C(=C1)C1=CC(C)=CC=C1C | |
210.32 | |
210.32 |
Chemical Identifiers
3075-84-1 | |
210.32 | |
ZHTROMYSDSTCCE-UHFFFAOYSA-N | |
2-(2,5-dimethylphenyl)-1,4-dimethylbenzene |
C16H18 | |
MFCD00151846 | |
137818 | |
CC1=CC=C(C)C(=C1)C1=CC(C)=CC=C1C |
Safety and Handling
EINECSNumber : 000-000-0
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only