missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,2,5,5-Tetramethyl-3-pyrrolin-1-oxyl-3-carboxylic Acid 99%, ACROS Organics™

$311.17 - $311.17

Chemical Identifiers

CAS 2154-67-8
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Molecular Formula C9H14NO3
Formula Weight 184.22
MDL Number MFCD00041847
Synonym 3-Carboxy-2,2,5,5-tetramethyl-3-pyrrolin-1-yloxy
Chemical Name or Material 2,2,5,5-Tetramethyl-3-pyrrolin-1-oxyl-3-carboxylic acid, 99%
Assay ≥98.5%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC420640010 SDS Fisher Scientific Acros
420640010
Glass bottle 1g Each for $311.17
AC420640050 SDS Fisher Scientific Acros
420640050
Glass bottle 5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

2154-67-8
100.0
C9H14NO3
MFCD00041847
2,2,5,5-Tetramethyl-3-pyrrolin-1-oxyl-3-carboxylic acid, 99%
98.5
99%
184.22
3-Carboxy-2,2,5,5-tetramethyl-3-pyrrolin-1-yloxy
≥98.5%

Specifications

Yellow
2154-67-8
100.0
Glass bottle
99%
184.22
3-Carboxy-2,2,5,5-tetramethyl-3-pyrrolin-1-yloxy
≥98.5%
1g
98.5
Authentic
Crystalline Powder
C9H14NO3
MFCD00041847
2,2,5,5-Tetramethyl-3-pyrrolin-1-oxyl-3-carboxylic acid, 99%

Safety and Handling

EINECSNumber : 218-447-7

TSCA : TSCA