missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,2,4-Trimethylpentane, HPLC Grade, >99.7%, Alfa Aesar™

Maximum level of impurities: Evaporation residue 3ppm, Chlorinated pesticides 5ppt, H2O 0.02%

Manufacturer:  Alfa Aesar 22901K7

 View more versions of this product

Catalog No. AA22901K7



Specifications

Specifications

Colorless liquid
410°C (770°F)
Colorless
540-84-1
C8H18
isooctane, isobutyltrimethylmethane, pentane, 2,2,4-trimethyl, iso-octane, 2,4,4-trimethylpentane, unii-qab8f5669o, ch3 2chch2c ch3 3, 2, 2, 4-trimethylpentane, i-octane, isobutyltrimethylethane
CC(C)CC(C)(C)C
114.232
CHEBI:62805
Liquid
2, 2, 4-Trimethylpentane
Nearly odorless
4L
>99.7%
MFCD00008943
NHTMVDHEPJAVLT-UHFFFAOYSA-N
2,2,4-trimethylpentane
10907
114.23
HPLC

Chemical Identifiers

540-84-1
114.232
NHTMVDHEPJAVLT-UHFFFAOYSA-N
10907
2,2,4-trimethylpentane
C8H18
MFCD00008943
isooctane, isobutyltrimethylmethane, pentane, 2,2,4-trimethyl, iso-octane, 2,4,4-trimethylpentane, unii-qab8f5669o, ch3 2chch2c ch3 3, 2, 2, 4-trimethylpentane, i-octane, isobutyltrimethylethane
CHEBI:62805
CC(C)CC(C)(C)C
Safety and Handling

Safety and Handling

H225-H304-H400-H410-H315-H336: Highly flammable liquid and vapour. May be fatal if swallowed and enters airways. Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects. Causes skin irritation. May cause drowsiness or dizziness.

DOTInformation : Hazard Class: 3, Packing Group: II

EINECSNumber : 208-759-1

SDS Documents