Please login to your online account to display your discounted pricing

2,2,4-Trimethylpentane, EMD Millipore


Quantity 4L
Packaging Glass Bottle

Manufacturer: emd millipore corporation  TX13891

 View more versions of this product

Catalog No. MTX13891

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 114.23
Quantity 4L
Density 0.69g/cm3 (20°C)
Molecular Formula C8H18
Synonym 2, 2, 4-Trimethylpentane
Ignition Point 410°C
Evaporation Number 2.3
Chemical Name or Material 2, 2, 4-Trimethylpentane
Vapor Pressure 51 hPa (20°C)
Packaging Glass Bottle
Saturation Concentration 239g/m3 (20°C)
Explosion Limit 1 to 5.5%(V)
Name Note SeccoSolv™, Dried
CAS 540-84-1
Recommended Storage Store at +15°C to +25°C
Linear Formula CH3C(CH3)2CH2CH(CH3)CH3

Specifications

Chemical Name or Material 2, 2, 4-Trimethylpentane
Name Note SeccoSolv™, Dried
CAS 540-84-1
Molecular Formula C8H18
Linear Formula CH3C(CH3)2CH2CH(CH3)CH3
Formula Weight 114.23
Synonym 2, 2, 4-Trimethylpentane
Accessories

Store at +15°C to +25°C