Please login to your online account to display your discounted pricing

2,2,4-Trimethylpentane, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

High quality chemicals for laboratory and specialized industrial use

$255.85 - $905.80

Chemical Identifiers

Chemical Name or Material 2,2,4-Trimethylpentane
CAS 540-84-1
Purity Grade Baker Analyzed™ ACS Reagent
Molecular Formula (CH3)3CCH2CH(CH3)2
Formula Weight 114.23
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02002060

 
avantor performance
947803
Narrow Mouth Amber Glass Bottle 4L Each for $905.80

02002063

 
avantor performance
947801
Narrow Mouth Amber Glass Bottle 500mL Each for $255.85
Description & Specifications

Specifications

Purity Grade Baker Analyzed™ ACS Reagent
Formula Weight 114.23
Chemical Name or Material 2,2,4-Trimethylpentane
Packaging Narrow Mouth Amber Glass Bottle
Molecular Formula (CH3)3CCH2CH(CH3)2
CAS 540-84-1
Flash Point −12.00°C
Specific Gravity 1L = 0.69kg
Quantity 4L