Please login to your online account to display your discounted pricing

2,2,4-Trimethylpentane, 99+%, pure, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2, 2, 4-Trimethylpentane
Name Note 99+%
CAS 540-84-1
CAS Min % 99.0
CAS Max % 100.0
Purity Grade Pure
Assay ≥99.0%
Assay Percent Range 99+%
Molecular Formula C8H18
Linear Formula (CH3)2CHCH2C(CH3)3
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC421975000

  SDS
thermo fisher scientific
421975000
Glass bottle 500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥99.0%
Physical Form Liquid
Formula Weight 114.23
Quantity 500g
Color Clear colorless
EINECS Number 208-759-1
TSCA TSCA
Purity Grade Pure
Density 0.6900g/mL
RTECS Number SA3320000
Beilstein 01, IV, 439
Merck Index 15, 5239
CAS Min % 99.0
CAS Max % 100.0
Refractive Index 1.3900 to 1.3920 (20°C, 589nm)
Chemical Name or Material 2, 2, 4-Trimethylpentane
Infrared Spectrum Authentic
Packaging Glass bottle
Name Note 99+%
Linear Formula (CH3)2CHCH2C(CH3)3
Synonym Isooctane
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
May be fatal if swallowed and enters airways.
Very toxic to aquatic life with long lasting effects.
May cause drowsiness or dizziness.
Causes skin irritation.
Highly flammable liquid and vapor.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Assay Percent Range 99+%
Molecular Formula C8H18
MDL Number MFCD00008943
CAS 540-84-1
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May be fatal if swallowed and enters airways.
Very toxic to aquatic life with long lasting effects.
May cause drowsiness or dizziness.
Causes skin irritation.
Highly flammable liquid and vapor.