Please login to your online account to display your discounted pricing

2,2,4-Trimethylpentane, 99+%, pure, ACROS Organics™


Quantity 500g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  421975000

 View more versions of this product

Catalog No. AC421975000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥99.0%
Physical Form Liquid
Formula Weight 114.23
Quantity 500g
Color Clear colorless
TSCA TSCA
RTECS Number SA3320000
EINECS Number 208-759-1
Density 0.6900g/mL
Purity Grade Pure
Beilstein 01, IV, 439
Merck Index 15, 5239
CAS Max % 100.0
CAS Min % 99.0
Refractive Index 1.3900 to 1.3920 (20°C, 589nm)
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Name Note 99+%
Packaging Glass bottle
Synonym Isooctane
Infrared Spectrum Authentic
Health Hazard 1 GHS Signal Word: Danger
Linear Formula (CH3)2CHCH2C(CH3)3
Chemical Name or Material 2, 2, 4-Trimethylpentane
Health Hazard 2 GHS H Statement:
May be fatal if swallowed and enters airways.
Very toxic to aquatic life with long lasting effects.
May cause drowsiness or dizziness.
Causes skin irritation.
Highly flammable liquid and vapor.
Molecular Formula C8H18
MDL Number MFCD00008943
CAS 540-84-1
Assay Percent Range 99+%

Specifications

Chemical Name or Material 2, 2, 4-Trimethylpentane
Name Note 99+%
CAS 540-84-1
CAS Min % 99.0
CAS Max % 100.0
Purity Grade Pure
Assay ≥99.0%
Assay Percent Range 99+%
Molecular Formula C8H18
Linear Formula (CH3)2CHCH2C(CH3)3
Formula Weight 114.23
MDL Number MFCD00008943
Synonym Isooctane
Accessories

GHS Signal Word: Danger

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May be fatal if swallowed and enters airways.
Very toxic to aquatic life with long lasting effects.
May cause drowsiness or dizziness.
Causes skin irritation.
Highly flammable liquid and vapor.

SDS & Safety Documents