Please login to your online account to display your discounted pricing

2,2,4-Trimethylpentane, 99.5%, for analysis, ACROS Organics™


Quantity 5mL
Packaging Glass bottle

Manufacturer: thermo fisher scientific  265440050

 View more versions of this product

Catalog No. AC265440050

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Liquid
Assay ≥99.5%
Formula Weight 114.23
Quantity 5mL
Color Clear
Beilstein 01, IV, 439
Merck Index 15, 5239
Purity Grade For analysis
Density 0.69g/mL
Residue after Evaporation 0.001% max.
Water 0.01% max. (Coulometric)
Packaging Glass bottle
Aluminum (Al) 0.5ppm max.
Assay Percent Range 99.50%
Linear Formula (CH3)2CHCH2C(CH3)3
MDL Number MFCD00008943
Health Hazard 2 GHS H Statement:
May be fatal if swallowed and enters airways.
Very toxic to aquatic life with long lasting effects.
May cause drowsiness or dizziness.
Causes skin irritation.
Highly flammable liquid and vapor.
Copper (Cu) 0.02ppm max.
Lead (Pb) 0.1ppm max.
Cadmium (Cd) 0.05ppm max.
CAS Min % 99.4
CAS Max % 100.0
Chemical Name or Material 2, 2, 4-Trimethylpentane
Hydrogen Sulfide and Organic S cmpds 50ppm max.
Infrared Spectrum Authentic
Iron (Fe) 0.1ppm max.
Nickel (Ni) 0.02ppm max.
Trace Analysis 0.1ppm max.
Molecular Formula C8H18
CAS 540-84-1
Synonym Isooctane
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 2, 2, 4-Trimethylpentane
CAS 540-84-1
CAS Min % 99.4
CAS Max % 100.0
Purity Grade For analysis
Assay ≥99.5%
Assay Percent Range 99.50%
Molecular Formula C8H18
Linear Formula (CH3)2CHCH2C(CH3)3
Formula Weight 114.23
MDL Number MFCD00008943
Synonym Isooctane
Accessories

GHS Signal Word: Danger

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May be fatal if swallowed and enters airways.
Very toxic to aquatic life with long lasting effects.
May cause drowsiness or dizziness.
Causes skin irritation.
Highly flammable liquid and vapor.

SDS & Safety Documents