Please login to your online account to display your discounted pricing

2,2,4-Trimethyl-1,3-pentanediolmono(2-methylpropanoate), +98%, ACROS Organics™


Quantity 500g
Packaging Glass bottle

Manufacturer: fisher scientific acros  422015000

 View more versions of this product

Catalog No. AC422015000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98.0% (sum two isomers)
Physical Form Liquid
Formula Weight 216.32
Quantity 500g
Color Clear
TSCA TSCA
RTECS Number UF6000000
EINECS Number 246-771-9
Density 0.9500g/mL
CAS Max % 100.0
CAS Min % 98.0
Packaging Glass bottle
Assay Percent Range 98+%
Synonym 2,2,4-Trimethyl-1,3-pentanediol monoisobutyrate
Refractive Index 1.4410 to 1.4430 (20°C, 589nm)
Infrared Spectrum Authentic
CAS 25265-77-4
MDL Number MFCD00148967
Chemical Name or Material 2,2,4-Trimethyl-1,3-pentanediolmono(2-methylpropanoate)
Molecular Formula C12H24O3

Specifications

Chemical Name or Material 2,2,4-Trimethyl-1,3-pentanediolmono(2-methylpropanoate)
CAS 25265-77-4
CAS Min % 98.0
CAS Max % 100.0
Assay ≥98.0% (sum two isomers)
Assay Percent Range 98+%
Molecular Formula C12H24O3
Formula Weight 216.32
MDL Number MFCD00148967
Synonym 2,2,4-Trimethyl-1,3-pentanediol monoisobutyrate