Please login to your online account to display your discounted pricing

2,2,4-Trimethyl-1,3-pentanediol Diisobutyrate 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2,2,4-Trimethyl-1,3-pentanediol diisobutyrate
Name Note 98%
CAS 6846-50-0
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C16H30O4
Formula Weight 286.4
MDL Number MFCD00059267
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC422000010

  SDS
fisher scientific acros
422000010
Glass bottle 1kg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC422000050

  SDS
thermo fisher scientific
422000050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Physical Form Liquid
Formula Weight 286.4
Quantity 1kg
Color Clear colorless
Merck Index 15, 9399
EINECS Number 229-934-9
TSCA TSCA
RTECS Number SA1420000
Density 0.94g/mL
CAS Max % 100.0
Chemical Name or Material 2,2,4-Trimethyl-1,3-pentanediol diisobutyrate
Packaging Glass bottle
Refractive Index 1.4330 to 1.4350 (20°C, 589nm)
Name Note 98%
Assay Percent Range 98%
CAS 6846-50-0
CAS Min % 97.5
Infrared Spectrum Authentic
Molecular Formula C16H30O4
MDL Number MFCD00059267