Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,2,4-Trimethyl-1,3-pentanediol, 94%

$50.62 - $195.88

Chemical Identifiers

Chemical Name or Material 2,2,4-Trimethyl-1,3-pentanediol
CAS 144-19-4
Molecular Formula C8H18O2
Formula Weight 146.23
MDL Number MFCD00004681
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2132809

  SDS
alfa aesar™
B2132809
10g Each for $50.62

AAB2132818

  SDS
alfa aesar™
B2132818
50g Each for $195.88
Description & Specifications

Specifications

Formula Weight 146.23
Melting Point 50° to 53°C
Assay 94%
MDL Number MFCD00004681
Quantity 10g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302
Harmful if swallowed.
CAS 144-19-4
Boiling Point 232°C
TSCA TSCA
Flash Point >110°C (230°F)
Molecular Formula C8H18O2
Health Hazard 3 GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material 2,2,4-Trimethyl-1,3-pentanediol
Safety and Handling

Warning

GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302
Harmful if swallowed.