missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2,2',4'-Trichloroacetophenone 98.0+%, TCI America™
Supplier: TCI America T1405250G
Specifications
2,2′,4′-Trichloroacetophenone | |
White | |
MFCD00000934 | |
1759 | |
VYWPPRLJNVHPEU-UHFFFAOYSA-N | |
2-chloro-1-(2,4-dichlorophenyl)ethanone | |
20250 | |
≥98.0% (GC) |
4252-78-2 | |
C8H5Cl3O | |
250 g | |
2,2',4'-trichloroacetophenone, 2-chloro-1-2,4-dichlorophenyl ethanone, 2,4-dichlorophenacyl chloride, unii-2e35p7hji2, 2-chloro-1-2,4-dichlorophenyl ethan-1-one, ccris 633, ethanone, 2-chloro-1-2,4-dichlorophenyl, 2,2,4-trichloroacetophenone, dsstox_cid_6190, 1-2,4-dichlorophenyl-2-chloroethan-1-one | |
C1=CC(=C(C=C1Cl)Cl)C(=O)CCl | |
223.477 | |
223.48 | |
Crystalline Powder |
Chemical Identifiers
4252-78-2 | |
223.477 | |
VYWPPRLJNVHPEU-UHFFFAOYSA-N | |
20250 | |
C1=CC(=C(C=C1Cl)Cl)C(=O)CCl |
C8H5Cl3O | |
MFCD00000934 | |
2,2',4'-trichloroacetophenone, 2-chloro-1-2,4-dichlorophenyl ethanone, 2,4-dichlorophenacyl chloride, unii-2e35p7hji2, 2-chloro-1-2,4-dichlorophenyl ethan-1-one, ccris 633, ethanone, 2-chloro-1-2,4-dichlorophenyl, 2,2,4-trichloroacetophenone, dsstox_cid_6190, 1-2,4-dichlorophenyl-2-chloroethan-1-one | |
2-chloro-1-(2,4-dichlorophenyl)ethanone |
Safety and Handling
TSCA : No