Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(2,4-Difluorophenoxy)-5-fluoro-N-methylbenzylamine, 96%

$76.39 - $76.39

Chemical Identifiers

Chemical Name or Material 2-(2,4-Difluorophenoxy)-5-fluoro-N-methylbenzylamine
Molecular Formula C14H12F3NO
Formula Weight 267.25
MDL Number MFCD08064232
Synonym 1-[2-(2,4-Difluorophenoxy)-5-fluorophenyl]-N-methylmethylamine
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AAH5072803

  SDS
alfa aesar™
H5072803
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH50728MD

  SDS
alfa aesar™
H50728MD
Each for $76.39
Description & Specifications

Specifications

Formula Weight 267.25
Quantity 1g
Synonym 1-[2-(2,4-Difluorophenoxy)-5-fluorophenyl]-N-methylmethylamine
MDL Number MFCD08064232
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Assay 96%
Molecular Formula C14H12F3NO
Health Hazard 1 Warning
Chemical Name or Material 2-(2,4-Difluorophenoxy)-5-fluoro-N-methylbenzylamine
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.