Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(2,4-Dichlorophenoxy)thioacetamide, 97%

$73.89 - $183.18

Chemical Identifiers

Chemical Name or Material 2-(2,4-Dichlorophenoxy)thioacetamide
CAS 2302-32-1
Molecular Formula C8H7Cl2NOS
Formula Weight 236.11
MDL Number MFCD00172128
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH26583MD

  SDS
alfa aesar™
H26583MD
250mg Each for $73.89

AAH2658303

  SDS
alfa aesar™
H2658303
1g Each for $183.18
Description & Specifications

Specifications

Formula Weight 236.11
Chemical Name or Material 2-(2,4-Dichlorophenoxy)thioacetamide
Assay 97%
Molecular Formula C8H7Cl2NOS
MDL Number MFCD00172128
CAS 2302-32-1
Melting Point 136° to 138°C
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 250mg
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.