Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(2,4-Dichlorophenoxy)propionic acid hydrazide, 98%

$33.59 - $446.00

Chemical Identifiers

Chemical Name or Material 2-(2,4-Dichlorophenoxy)propionic acid hydrazide
CAS 15253-89-1
Molecular Formula C9H10Cl2N2O2
Formula Weight 249.09
MDL Number MFCD00204152
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1681703

 
alfa aesar™
A1681703
1g Each for $33.59

AAA1681714

 
alfa aesar™
A1681714
25g Each for $446.00
Description & Specifications

Specifications

Formula Weight 249.09
Assay 98%
Molecular Formula C9H10Cl2N2O2
Chemical Name or Material 2-(2,4-Dichlorophenoxy)propionic acid hydrazide
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 1 Warning
CAS 15253-89-1
MDL Number MFCD00204152
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.