Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(2,4-Dichlorophenoxy)aniline hydrochloride, 98%

$78.04 - $323.24

Chemical Identifiers

Chemical Name or Material 2-(2,4-Dichlorophenoxy)aniline hydrochloride
CAS 89279-16-3
Molecular Formula C12H9Cl2NO·HCl
Formula Weight 290.58
MDL Number MFCD00052001
Beilstein 3736700
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2034506

 
alfa aesar™
B2034506
5g Each for $78.04

AAB2034514

 
alfa aesar™
B2034514
25g Each for $323.24
Description & Specifications

Specifications

Formula Weight 290.58
Chemical Name or Material 2-(2,4-Dichlorophenoxy)aniline hydrochloride
Assay 98%
Molecular Formula C12H9Cl2NO·HCl
MDL Number MFCD00052001
CAS 89279-16-3
Beilstein 3736700
Melting Point 96° to 98°C
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 5g
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.