Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(2,4,5-Trifluorophenyl)ethanol, 97%

$57.11 - $188.35

Chemical Identifiers

Chemical Name or Material 2-(2,4,5-Trifluorophenyl)ethanol
CAS 883267-70-7
Molecular Formula C8H7F3O
Formula Weight 176.14
MDL Number MFCD06660356
Synonym 2,4,5-Trifluorophenethyl alcohol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3268603

  SDS
alfa aesar™
H3268603
1g Each for $188.35

AAH32686MD

  SDS
alfa aesar™
H32686MD
250mg Each for $57.11
Description & Specifications

Specifications

Formula Weight 176.14
Molecular Formula C8H7F3O
Synonym 2,4,5-Trifluorophenethyl alcohol
MDL Number MFCD06660356
Quantity 1g
Chemical Name or Material 2-(2,4,5-Trifluorophenyl)ethanol
Assay 97%
CAS 883267-70-7