Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(2,4,5-Trichlorophenoxy)propionic acid, 96%

$108.07 - $108.07

Chemical Identifiers

Chemical Name or Material 2-(2,4,5-Trichlorophenoxy)propionic acid
CAS 93-72-1
Molecular Formula C9H7Cl3O3
Formula Weight 269.51
MDL Number MFCD00002646
UN Number UN3077
Beilstein 1985768
Synonym Fenoprop; Silvex
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0813606

  SDS
alfa aesar™
L0813606*
5g Each for $108.07

AAL0813614

  SDS
alfa aesar™
L0813614
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 269.51
Chemical Name or Material 2-(2,4,5-Trichlorophenoxy)propionic acid
Assay 96%
Molecular Formula C9H7Cl3O3
MDL Number MFCD00002646
CAS 93-72-1
UN Number UN3077
Beilstein 1985768
Synonym Fenoprop; Silvex
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302-H315
Harmful if swallowed.
Causes skin irritation.
Health Hazard 3 GHS P Statement
P280-P362-P301+P312-P321-P332+P313-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
Take off contaminated clothing and wash before reuse.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 176° to 180°C
DOT Information Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
RTECS Number UF8225000
Quantity 5g
Safety and Handling

Warning

GHS P Statement
P280-P362-P301+P312-P321-P332+P313-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
Take off contaminated clothing and wash before reuse.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H315
Harmful if swallowed.
Causes skin irritation.