missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,2',4,4'-Tetrahydroxybenzophenone, 98+%

Manufacturer:  Alfa Aesar™ A1023714

 View more versions of this product

Catalog No. AAA1023714



Description

Description

Applications
2,2′,4,4′-Tetrahydroxybenzophenone is used as the raw material of ultraviolet absorbers in cosmetics, plastics, resins and rubber. It is used as pharmaceutical intermediates and light-sensitive materials. It is also used as pharmaceutical adjuvant.

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Store in a cool, dry condition in well sealed containers.
Specifications

Specifications

2,2',4,4'-Tetrahydroxybenzophenone
25g
C13H10O5
1914746
WXNRYSGJLQFHBR-UHFFFAOYSA-N
bis(2,4-dihydroxyphenyl)methanone
8571
≥98%
198°C to 203°C
131-55-5
MFCD00002278
2,2',4,4'-tetrahydroxybenzophenone, bis 2,4-dihydroxyphenyl methanone, benzophenone-2, uvinol d-50, uvinul d-50, methanone, bis 2,4-dihydroxyphenyl, 2,4,2',4'-tetrahydroxybenzophenone, unii-prr8k3h9vn, 2,2',4,4'-tetrehydroxybenzophenone, prr8k3h9vn
C1=CC(=C(C=C1O)O)C(=O)C2=C(C=C(C=C2)O)O
246.218
246.22

Chemical Identifiers

131-55-5
246.218
WXNRYSGJLQFHBR-UHFFFAOYSA-N
8571
C1=CC(=C(C=C1O)O)C(=O)C2=C(C=C(C=C2)O)O
C13H10O5
MFCD00002278
2,2',4,4'-tetrahydroxybenzophenone, bis 2,4-dihydroxyphenyl methanone, benzophenone-2, uvinol d-50, uvinul d-50, methanone, bis 2,4-dihydroxyphenyl, 2,4,2',4'-tetrahydroxybenzophenone, unii-prr8k3h9vn, 2,2',4,4'-tetrehydroxybenzophenone, prr8k3h9vn
bis(2,4-dihydroxyphenyl)methanone
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

RTECSNumber : DJ1892000

TSCA : TSCA

SDS Documents