missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,2',4,4'-Tetrahydroxybenzophenone, 98+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1023722

 View more versions of this product

Catalog No. AAA1023722



Description

Description

2,2',4,4'-Tetrahydroxybenzophenone is used as the raw material of ultraviolet absorbers in cosmetics, plastics, resins and rubber. It is used as pharmaceutical intermediates and light-sensitive materials. It is also used as pharmaceutical adjuvant.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,2′,4,4′-Tetrahydroxybenzophenone is used as the raw material of ultraviolet absorbers in cosmetics, plastics, resins and rubber. It is used as pharmaceutical intermediates and light-sensitive materials. It is also used as pharmaceutical adjuvant.

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Store in a cool, dry condition in well sealed containers.
Specifications

Specifications

2,2',4,4'-Tetrahydroxybenzophenone
198°C to 203°C
C13H10O5
100 g
2,2',4,4'-tetrahydroxybenzophenone, bis 2,4-dihydroxyphenyl methanone, benzophenone-2, uvinol d-50, uvinul d-50, methanone, bis 2,4-dihydroxyphenyl, 2,4,2',4'-tetrahydroxybenzophenone, unii-prr8k3h9vn, 2,2',4,4'-tetrehydroxybenzophenone, prr8k3h9vn
WXNRYSGJLQFHBR-UHFFFAOYSA-N
bis(2,4-dihydroxyphenyl)methanone
8571
≥98%
131-55-5
1.216
MFCD00002278
1914746
Slightly soluble in water.
C1=CC(=C(C=C1O)O)C(=O)C2=C(C=C(C=C2)O)O
246.218
246.22

Chemical Identifiers

131-55-5
246.218
WXNRYSGJLQFHBR-UHFFFAOYSA-N
8571
C1=CC(=C(C=C1O)O)C(=O)C2=C(C=C(C=C2)O)O
C13H10O5
MFCD00002278
2,2',4,4'-tetrahydroxybenzophenone, bis 2,4-dihydroxyphenyl methanone, benzophenone-2, uvinol d-50, uvinul d-50, methanone, bis 2,4-dihydroxyphenyl, 2,4,2',4'-tetrahydroxybenzophenone, unii-prr8k3h9vn, 2,2',4,4'-tetrehydroxybenzophenone, prr8k3h9vn
bis(2,4-dihydroxyphenyl)methanone
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

EINECSNumber : 205-028-9

RTECSNumber : DJ1892000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only