Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,2,4,4-Tetrafluoro-1,3-dithietane, 97%

Chemical Identifiers

Chemical Name or Material 2,2,4,4-Tetrafluoro-1,3-dithietane
CAS 1717-50-6
Molecular Formula C2F4S2
Formula Weight 164.14
MDL Number MFCD00053538
UN Number UN1993
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2162009

  SDS
alfa aesar™
B2162009
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2162018

  SDS
alfa aesar™
B2162018
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 164.14
Chemical Name or Material 2,2,4,4-Tetrafluoro-1,3-dithietane
Assay 97%
Molecular Formula C2F4S2
MDL Number MFCD00053538
CAS 1717-50-6
UN Number UN1993
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H226
Flammable liquid and vapour.
Health Hazard 3 GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Boiling Point 47° to 49°C
Flash Point 36°C (96°F)
Odor Unpleasant
Quantity 10g
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
Safety and Handling

Warning

GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H226
Flammable liquid and vapour.