missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2,2'-[4-(2-Hydroxyethylamino)-3-nitrophenylimino]diethanol 98.0+%, TCI America™
Supplier: TCI America T278725G
Specifications
2,2′-[4-(2-Hydroxyethylamino)-3-nitrophenylimino]diethanol | |
108°C | |
MFCD00071762 | |
N1,N4,N4-Tris(2-hydroxyethyl)-2-nitro-1,4-phenylenediamine | |
OCCNC1=CC=C(C=C1[N+]([O-])=O)N(CCO)CCO | |
285.30 | |
CHEBI:82472 | |
≥98.0% (HPLC,T) |
33229-34-4 | |
C12H19N3O5 | |
25 g | |
MIWUTEVJIISHCP-UHFFFAOYSA-N | |
2-({4-[bis(2-hydroxyethyl)amino]-2-nitrophenyl}amino)ethan-1-ol | |
36383 | |
285.30 | |
Crystalline Powder |
Chemical Identifiers
33229-34-4 | |
285.30 | |
MIWUTEVJIISHCP-UHFFFAOYSA-N | |
36383 | |
2-({4-[bis(2-hydroxyethyl)amino]-2-nitrophenyl}amino)ethan-1-ol |
C12H19N3O5 | |
MFCD00071762 | |
N1,N4,N4-Tris(2-hydroxyethyl)-2-nitro-1,4-phenylenediamine | |
CHEBI:82472 | |
OCCNC1=CC=C(C=C1[N+]([O-])=O)N(CCO)CCO |
Safety and Handling
RTECSNumber : KL2880000
TSCA : No