Please login to your online account to display your discounted pricing

2,2,3-Trimethylbutane, 99%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC398620010

 View more versions of this product

Catalog No. AC398620010

  • / Each
 

Description & Specifications

Chemical Identifiers

Purity Grade Extra Pure
Physical Form Liquid
Assay ≥98.5%
Formula Weight 100.2
Quantity 1g
Color Clear colorless
Density 0.69g/mL
CAS Max % 100.0
Chemical Name or Material 2, 2, 3-Trimethylbutane
Packaging Glass bottle
Molecular Formula C7H16
CAS 464-06-2
Infrared Spectrum Authentic
Refractive Index 1.3880 to 1.3900 (20°C, 589nm)
Assay Percent Range 99%
Linear Formula CH3C(CH3)2CH(CH3)2
MDL Number MFCD00039846
CAS Min % 98.5
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
May cause drowsiness or dizziness.
Very toxic to aquatic life with long lasting effects.
May be fatal if swallowed and enters airways.
Causes skin irritation.
Highly flammable liquid and vapor.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 2, 2, 3-Trimethylbutane
CAS 464-06-2
CAS Min % 98.5
CAS Max % 100.0
Purity Grade Extra Pure
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C7H16
Linear Formula CH3C(CH3)2CH(CH3)2
Formula Weight 100.2
MDL Number MFCD00039846
Accessories

GHS Signal Word: Danger

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause drowsiness or dizziness.
Very toxic to aquatic life with long lasting effects.
May be fatal if swallowed and enters airways.
Causes skin irritation.
Highly flammable liquid and vapor.