Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(2,3-Difluorophenyl)ethanol, 97%

$56.50 - $187.80

Chemical Identifiers

Chemical Name or Material 2-(2,3-Difluorophenyl)ethanol
CAS 126163-30-2
Molecular Formula C8H8F2O
Formula Weight 158.15
MDL Number MFCD01862011
Synonym 2,3-Difluorophenethyl alcohol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3240803

  SDS
alfa aesar™
H3240803
1g Each for $187.80

AAH32408MD

  SDS
alfa aesar™
H32408MD
250mg Each for $56.50
Description & Specifications

Specifications

Formula Weight 158.15
Chemical Name or Material 2-(2,3-Difluorophenyl)ethanol
Assay 97%
Molecular Formula C8H8F2O
MDL Number MFCD01862011
CAS 126163-30-2
Synonym 2,3-Difluorophenethyl alcohol
Quantity 1g