missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,2,3,4,4,4-Hexafluorobutyl methacrylate, 96%, stab.

Manufacturer:  Alfa Aesar™ L0954306

 View more versions of this product

Catalog No. AAL0954306



Description

Description

Applications
It is an epoxy thinner and is mainly used as the modifier of the (meth)acrylate resin. Well-defined organic/inorganic hybrid fluorinated star polymers were synthesized via atom transfer radical polymerization (ATRP) of 2,2,3,4,4,4-hexafluorobutyl methacrylate (HFBMA) using octa(aminophenyl)silsesquioxane (OAPS) nano-cage as initiator.

Solubility
Difficult to mix in water.

Notes
Store at room temperature. Incompatible with strong oxidizing agents.
Specifications

Specifications

2,2,3,4,4,4-Hexafluorobutyl methacrylate
158°C
stabilized
C8H8F6O2
UN3272
2,2,3,4,4,4-hexafluorobutyl methacrylate, 1h,1h,3h-hexafluorobutyl methacrylate, hexafluorobutyl methacrylate, methacrylic acid 2,2,3,4,4,4-hexafluorobutyl ester, 2-propenoic acid, 2-methyl-, 2,2,3,4,4,4-hexafluorobutyl ester, hexafluorobutyl methacrylate polymer, hfbma, acmc-1adv6, ksc489s1j, 2-propenoic acid, 2-methyl-, 2,2,3,4,4,4-hexafluorobutyl ester, homopolymer
CC(=C)C(=O)OCC(C(C(F)(F)F)F)(F)F
250.14
250.14
56°C (132°F)
5g
36405-47-7
MFCD00042311
2725177
DFVPUWGVOPDJTC-UHFFFAOYSA-N
2,2,3,4,4,4-hexafluorobutyl 2-methylprop-2-enoate
549772
96%

Chemical Identifiers

36405-47-7
250.14
DFVPUWGVOPDJTC-UHFFFAOYSA-N
549772
CC(=C)C(=O)OCC(C(C(F)(F)F)F)(F)F
C8H8F6O2
MFCD00042311
2,2,3,4,4,4-hexafluorobutyl methacrylate, 1h,1h,3h-hexafluorobutyl methacrylate, hexafluorobutyl methacrylate, methacrylic acid 2,2,3,4,4,4-hexafluorobutyl ester, 2-propenoic acid, 2-methyl-, 2,2,3,4,4,4-hexafluorobutyl ester, hexafluorobutyl methacrylate polymer, hfbma, acmc-1adv6, ksc489s1j, 2-propenoic acid, 2-methyl-, 2,2,3,4,4,4-hexafluorobutyl ester, homopolymer
2,2,3,4,4,4-hexafluorobutyl 2-methylprop-2-enoate
Safety and Handling

Safety and Handling

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P305+P351+P338-P302+P352
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN: Wash with plenty of soap and water.

Warning

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ESTERS, N.O.S.

Recommended Storage : 2° to 8°C

Documents