missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,2,3,4,4,4-Hexafluorobutyl Acrylate (stabilized with TBC), TCI America™

$73.33 - $253.61

Chemical Identifiers

CAS 54052-90-3
Molecular Formula C7H6F6O2
Molecular Weight (g/mol) 236.113
MDL Number MFCD00044091
InChI Key LMVLEDTVXAGBJV-UHFFFAOYSA-N
Synonym Acrylic Acid 2,2,3,4,4,4-Hexafluorobutyl Ester, HFBA
PubChem CID 2774977
IUPAC Name 2,2,3,4,4,4-hexafluorobutyl prop-2-enoate
SMILES C=CC(=O)OCC(C(C(F)(F)F)F)(F)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
H155425G View Documents TCI America
H155425G
25g Each for $253.61
 
H15545G View Documents TCI America
H15545G
5g Each for $73.33
 
Specifications

Chemical Identifiers

54052-90-3
236.113
LMVLEDTVXAGBJV-UHFFFAOYSA-N
2774977
C=CC(=O)OCC(C(C(F)(F)F)F)(F)F
C7H6F6O2
MFCD00044091
Acrylic Acid 2,2,3,4,4,4-Hexafluorobutyl Ester, HFBA
2,2,3,4,4,4-hexafluorobutyl prop-2-enoate

Specifications

43°C
25g
C7H6F6O2
3272
LMVLEDTVXAGBJV-UHFFFAOYSA-N
2,2,3,4,4,4-hexafluorobutyl prop-2-enoate
2774977
Clear Liquid at 20°C
2,2,3,4,4,4-Hexafluorobutyl Acrylate (stabilized with TBC)
Colorless
54052-90-3
MFCD00044091
Acrylic Acid 2,2,3,4,4,4-Hexafluorobutyl Ester, HFBA
C=CC(=O)OCC(C(C(F)(F)F)F)(F)F
236.113
236.11
≥98.0% (GC)
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator